missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115748
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PGP.
Glycerol-3-phosphate phosphatase hydrolyzing glycerol-3-phosphate into glycerol. Thereby, regulates the cellular levels of glycerol-3-phosphate a metabolic intermediate of glucose, lipid and energy metabolism. Was also shown to have a 2-phosphoglycolate phosphatase activity and a tyrosine-protein phosphatase activity. However, their physiological relevance is unclear. In vitro, has also a phosphatase activity toward ADP, ATP, GDP and GTP.
Specifications
| PGP | |
| Polyclonal | |
| Unconjugated | |
| Pgp | |
| 1700012G19Rik; AI481330; Aspartate-based ubiquitous Mg(2+)-dependent phosphatase; AUM; G3PP; Glycerol-3-phosphate phosphatase; PGP; PGPase; phosphoglycolate phosphatase; RGD1307773 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 283871, 287115, 67078 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| A6NDG6, D3ZDK7, Q8CHP8 | |
| Pgp | |
| A synthesized peptide derived from human PGP(Accession A6NDG6), corresponding to amino acid residues F176-A226. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction