missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGP9.5 (UCHL1) Monoclonal Antibody (OTI1A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA814089S
This item is not returnable.
View return policy
Description
PGP9.5 (UCHL1) Monoclonal antibody specifically detects PGP9.5 (UCHL1) in Human samples. It is validated for ELISASpecifications
| PGP9.5 (UCHL1) | |
| Monoclonal | |
| See Label | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P09936 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 7345 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| ELISA | |
| OTI1A8 | |
| Unconjugated | |
| Uchl1 | |
| AW822034; C88048; epididymis luminal protein 117; epididymis secretory protein Li 53; gad; gracile axonal dystrophy; HEL-117; HEL-S-53; MGC133697 protein; NDGOA; neuron cytoplasmic protein 9.5; OTTHUMP00000218137; OTTHUMP00000218139; OTTHUMP00000218140; OTTHUMP00000218141; PARK5; PGP 9.5; PGP9.5; PGP95; protein gene product 9.5; R75593; ubiquitin carboxyl-terminal esterase; ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase); ubiquitin carboxyl-terminal hydrolase isozyme L1; ubiquitin carboxyl-terminal hydrolase L1; ubiquitin carboxy-terminal hydrolase L1; ubiquitin C-terminal hydrolase; ubiquitin C-terminal hydrolase 1; ubiquitin C-terminal hydrolase L1; ubiquitin thioesterase L1; ubiquitin thiolesterase; ubiquitin thiolesterase L1; UCHL; UCHL1; UCH-L1; UCHL-1 | |
| Full length human recombinant protein of human UCHL1 (NP_004172) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction