Learn More
PGR Monoclonal Antibody (OTI11G9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA800666S
Encompass_Preferred
 View more versions of this product
Product Code. 17568732
  • € 242.00 / 30µL
Description
PGR Monoclonal antibody specifically detects PGR in Human samples. It is validated for Western Blot
Specifications
Specifications
PGR
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9H160
Mouse
Affinity chromatography
RUO
3622
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Western Blot
OTI11G9
Unconjugated
Ing2
9930019P03Rik; BB114106; ENSMUSG00000074510; LOW QUALITY PROTEIN: progesterone receptor; NR3C3; Nuclear receptor subfamily 3 group C member 3; p4 receptor; PGR; pot. alt. progesterone receptor; putative; PR; PR-A; PR-B; Progesterone receptor; progesterone receptor A; progesterone-receptor; putative form B; steroid hormone receptor; testicular progesterone receptor; unnamed protein product
Full length human recombinant protein of human ING2 produced in E.coli.