missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGR Monoclonal Antibody (OTI7G7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802712S
This item is not returnable.
View return policy
Description
PGR Monoclonal antibody specifically detects PGR in Human samples. It is validated for Western BlotSpecifications
| PGR | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P06401 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5241 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI7G7 | |
| Unconjugated | |
| PGR | |
| 9930019P03Rik; BB114106; ENSMUSG00000074510; LOW QUALITY PROTEIN: progesterone receptor; NR3C3; Nuclear receptor subfamily 3 group C member 3; p4 receptor; PGR; pot. alt. progesterone receptor; putative; PR; PR-A; PR-B; Progesterone receptor; progesterone receptor A; progesterone-receptor; putative form B; steroid hormone receptor; testicular progesterone receptor; unnamed protein product | |
| Human recombinant protein fragment corresponding to amino acids 1-298 of human PGR produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction