missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGR, Mouse anti-Human, Clone: 4E9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00005241-M08.100ug
This item is not returnable.
View return policy
Description
Sequence: MTELKAKGPRAPHVAGGPPSPEVGSPLLCRPAAGPFPGSQTSDTLPEVSAIPISLDGLLFPRPCQGQDPSDEKTQDQQSLSDVEGAYSRAEATRGAGGSSSSPPEKDSGLSpecifications
| PGR | |
| Monoclonal | |
| Unconjugated | |
| progesterone receptor | |
| NR3C3/PR | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 4E9 | |
| In 1x PBS, pH 7.4 | |
| NM_000926 | |
| PGR | |
| PGR (NP_000917, 1 a.a. ∼ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 5241 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction