missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGRMC1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36525AF647
This item is not returnable.
View return policy
Description
PGRMC1 Polyclonal antibody specifically detects PGRMC1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| PGRMC1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| HPR6.6PGRMC, membrane-associated progesterone receptor component 1, MPR, progesterone binding protein, progesterone receptor membrane component 1 | |
| The immunogen recognized by this antibody maps to a region between residue 145 to 195 of human Progesterone Receptor Membrane Component 1 using the numbering given in entry NP_006658.1. | |
| 0.1 mL | |
| Cancer | |
| 10857 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction