Learn More
PGRMC2 Mouse anti-Human, CoraLite 594, Clone: 5F10D7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL594-60249-100UL
Product Code. 17402653
  • € 450.00 / 100µL
Description
The PGRMC2 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human PGRMC2. This antibody recognizes human, mouse antigen. The PGRMC2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Specifications
PGRMC2
Monoclonal
CoraLite 594
100 μL
10424
IgG2a
Immunofluorescence
5F10D7