Learn More
PGRMC2 Mouse anti-Human, CoraLite 594, Clone: 5F10D7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-60249-100UL
Description
The PGRMC2 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human PGRMC2. This antibody recognizes human, mouse antigen. The PGRMC2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
| PGRMC2 | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 10424 | |
| IgG2a |
| Immunofluorescence | |
| 5F10D7 | |
| Mouse | |
| Primary | |
| Human, Mouse |