Learn More
pH buffer sol. NBS DIN 19226 pH 1.69, Mettler Toledo™
Product Code. 10517711 Shop All Mettler Toledo Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10517711 1 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10517711 Supplier Mettler Toledo Supplier No. 51319000
Specifications
Color Colorless
pH 1.69
Physical Form Liquid
Quantity 1 mL
Packaging 1Each
Solubility Information Fully miscible in water
compliance-icons
Product Identifier
  • Buffer pH 1.679 NBS MIC
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.