missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH buffer sol. NBS DIN 19226 pH 7.4, Mettler Toledo™
Shop All Mettler Toledo Products
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Specifications
Specifications
| Color | Colorless |
| pH | 7.4 |
| Physical Form | Liquid |
| Quantity | 250 mL |
| Packaging | 1Each |
| Solubility Information | Fully miscible in water |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction