Learn More
pH Buffer Standard Solutions pH 10.012, Hach Lange™
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- pH Standard, IUPAC, pH10.012 at 25 °C, Certified, 500 mL (Radiometer Analytical)
- 10.012 ± 0.010 pH at 25°C concentration
- Traceable to IUPAC pH scale and NIST
- Shelf life of 48 months from production date
- Each bottle (500 mL) is supplied with a COFRAC Verification Certificate and Certificate of Conformity and Traceability drawn up according to ISO31
- Total Control of the traceability chain and calculated uncertainties
- Direct traceability to primary standards and the reference method
- The competence and integrity of a laboratory internationally recognized to be accredited for the measurement of pH and conductivity reference materials
Specifications
Specifications
| Melting Point | 0°C |
| Color | Colorless |
| pH | 10.012 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Odor | Odorless |
| Traceability to NIST | Yes |