missing translation for 'onlineSavingsMsg'
Learn More

Buffer Solution, pH 4.00, Red, Chem-Lab

Product Code. 11913073
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11913073 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11913073 Supplier Chem-Lab Supplier No. CL03.0702.0100

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Designed for pH measurement and maintenance in moderately acidic samples. Buffer Solution, pH 4.00, Red, Chem-Lab is a solution of citrate, sodium chloride and sodium hydroxide, with a pH level accurate to ±0.02. It is color-coded for easy identification. The solution is traceable to SRM from NIST.

pH Levels ± 0.02:
  • pH 4.03 — 0°C
  • pH 4.02 — 10°C
  • pH 4.00 — 20°C
  • pH 4.00 — 25°C
  • pH 4.00 — 30°C
  • pH 4.00 — 40°C
  • pH 4.00 — 50°C
  • pH 4.00 — 60°C
  • pH 4.00 — 70°C
  • pH 4.00 — 80°C
  • pH 4.00 — 90°C

Specifications

Melting Point 0°C
Boiling Point 100°C
Color Red
Density 1.00g/mL
For Use With (Application) Laboratory use, pH measurements
pH 4.00 ±0.02
Physical Form Liquid
Quantity 100 mL
Solution Type pH Buffer
Formulation Citrate-NaCl-NaOH
Odor Odorless
Packaging Polyethylene
Traceability to NIST Yes
Solubility Information Soluble in water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.