Learn More
pH Buffers Solutions pH 6.00, Chem Lab™
Brand: Chem Lab CL03.0215.1000
Additional Details : Weight : 1.01000kg
Specifications
Colorless | |
6.00 | |
1000 mL | |
Odorless | |
Yes |
Laboratory use, pH measurements, traceable to SRM from NIST | |
Liquid | |
Citrate-NaOH | |
Polyethylene | |
Soluble in water |