Learn More
WTW™ pH Combination Electrode
Product Code. 10429192
Change view
Click to view available options
Material:
Epoxy or Polymer
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10429192 Epoxy or Polymer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10429192 Supplier WTW™ Supplier No. 109100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
The SensoLyt™ measuring chains without arming can be connected directly to a pH monitor pH 298 series. Depending on the application, you can install them into a flow vessel.
TRUSTED_SUSTAINABILITY
Specifications
Connector Types Watertight plug head connector
Type pH Combination Electrode
Filling Solution Type Gel Polymer solid electrolyte (AgCl-free)
Junction Hole junction
Material Epoxy or Polymer
pH Range 2 to 12
Temperature (Metric) Operating 0°C to 60°C