Learn More
SI Analytics™ pH Combination Electrode
Shop All SI Analytics Products
Click to view available options
Length (Metric):
120 mm
170 mm
320 mm
500 mm
600 mm
Junction:
Ceramic Diaphragm
Ground Joint Diaphragm
Platinum Diaphragm
Temperature (Metric) Range:
-5°C to +100°C
0°C to 100°C
0°C to 40°C
10°C to 40°C
Material:
A-Glass
H-Glass
Unit Size:
Each
Description
- Proven high-end electrodes for demanding measurement tasks
- Double junction Silamid™ reference system for fast and stable acquiring of measured values and for longer electrode life
- Utmost versatility of pH electrodes is achieved by a large selection of junctions, membrane glass types and shapes, shaft lengths and diameters, ground joints, plug connections and integrated temperature sensors
- Each pH combination electrode comes with individual serial number and quality certificate
Specifications
Specifications
| Connector Types | Plug Head |
| Junction | Ceramic Diaphragm |
| Material | H-Glass |
| pH Range | 0 to 14 |
| Type | Metal Combination Electrode |
| Internal Reference | Silamid™ |
| Membrane | Sphere |
| Model | Agcl 62 |
| Temperature (Metric) Range | 0°C to 100°C |
| Length (Metric) | 170 mm |
| Show More |