Learn More
SI Analytics™ pH Combination Electrode
Shop All SI Analytics Products
Click to view available options
Length (Metric):
120 mm
170 mm
320 mm
500 mm
600 mm
Junction:
Ceramic Diaphragm
Ground Joint Diaphragm
Platinum Diaphragm
Temperature (Metric) Range:
-5°C to +100°C
0°C to 100°C
0°C to 40°C
10°C to 40°C
Material:
A-Glass
H-Glass
Unit Size:
Each
Beskrivelse
- Proven high-end electrodes for demanding measurement tasks
- Double junction Silamid™ reference system for fast and stable acquiring of measured values and for longer electrode life
- Utmost versatility of pH electrodes is achieved by a large selection of junctions, membrane glass types and shapes, shaft lengths and diameters, ground joints, plug connections and integrated temperature sensors
- Each pH combination electrode comes with individual serial number and quality certificate
Tekniske data
Tekniske data
| Connector Types | Plug Head |
| Junction | Ceramic Diaphragm |
| Material | A-Glass |
| pH Range | 0 to 14 |
| Type | Combination pH Electrode |
| Internal Reference | Silamid™ |
| Membrane | Sphere |
| Model | N 6280 |
| Temperature (Metric) Range | -5°C to +100°C |
| Length (Metric) | 120 mm |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion