Learn More
Cole-Parmer™ pH Electrode Sealed Single-junction for Beckman Meters
Shop All Cole-Parmer Products
Click to view available options
For Use With (Equipment):
Beckman Meters
Unit Size:
Each
Description
- Electrodes designed to work with meters you already have.
- Seven different designs available—find the design that works best for your application without losing automatic temperature compensation.
- ''All-in-One'' electrodes offer versatility and customization.
- These electrodes combine the pH reference, pH half-cell, and temperature sensor in one 12 mm diameter electrode body.
- The seven designs allow you to select between low-maintenance, sealed electrodes or clog-resistant refillable electrodes.
- Tuff-Tip electrodes feature a semi-domed tip which protects the pH bulb and ensures a quick response in harsh and dirty samples.
- PBR-style electrodes provide predictable, reliable, and quick responses—no matter what the sample temperature.
- This design virtually eliminates drift, accuracy problems, and premature performance losses due to temperature.
- All electrodes include 3 ft. cable and BNC connector for pH, except where noted.
- Beckman Instruments-compatible electrodes feature a 30K temperature compensator with a 3.5 mm audio plug for connecting to your meter.
Specifications
Specifications
| Connector Types | BNC |
| Type | pH Electrode |
| Includes | 3 ft. Cable, BNC Connector |
| For Use With (Equipment) | Beckman Meters |