Learn More
Macherey Nagel pH Indicator Papers
The longstanding, appreciated standard for many pH testing applications.
Brand: Macherey Nagel 90214
This item is not returnable.
View return policy
Description
- pH indicator papers have been on the market for decades and are the appreciated standard for many applications
- For each pH value these papers show a single color which can be matched with the color scale at intervals of 0.2–1 pH unit
Specifications
| 12 to 14 | |
| pH Indicator Paper | |
| 7 mm |
| 1 x 5 m x 7 mm Spool on a Reel | |
| 5 m |