Learn More
Macherey Nagel pH Indicator Papers
The longstanding, appreciated standard for many pH testing applications.
Brand:  Macherey Nagel 90214
Product Code. 10303651
  • € 13.15 / Pack of 5
Only null left
In Stock
Description
  • pH indicator papers have been on the market for decades and are the appreciated standard for many applications
  • For each pH value these papers show a single color which can be matched with the color scale at intervals of 0.2–1 pH unit
Specifications
Specifications
12 to 14
pH Indicator Paper
7 mm
1 x 5 m x 7 mm Spool on a Reel
5 m