missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Blue Litmus Paper Reel

Provides a general test for acidic or alkaline reactions. Macherey Nagel™ Blue Litmus Paper Reels are comprised of completely impregnated indicator paper.

Brand:  Macherey-Nagel™ 91106

Additional Details : Weight : 0.01620kg

Product Code. 10373641

  • € 12.30 / 1 roll
Estimated Shipment: 08-05-2024
to see stock.
Add to basket

Description

Description

Provides a general test for acidic or alkaline reactions. Macherey Nagel™ Blue Litmus Paper Reels are comprised of completely impregnated indicator paper.

  • Color change from blue to red
  • Store in a cool, dry place
  • Avoid exposure to sunlight and moisture
  • Serves in neutralization reactions to check whether the change from acid to alkaline or vice versa has taken place
Specifications

Specifications

Blue to Red
Reel
1 Each
5000 mm
8 to 5
Determination of acidic and alkaline reaction
pH Paper
7 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers