Learn More
Macherey-Nagel™ pH Paper
Product Code. 10527341 Shop All Macherey-Nagel Products
Click to view available options
pH Range:
0.5 to 5.5
1 to 11
1 to 14
3.8 to 5.8
4 to 7
5.4 to 7
5.5 to 9
6.4 to 8
7.2 to 9.7
9 to 13
Unit Size:
5m
Each
Pack of 5
Product Code. 10527341
Brand: Macherey-Nagel™ 90213
Measure pH simply and easily. Macherey-Nagel™ pH paper comes in a circular format with a wide variety of pH ranges.
  • Universal and special indicator papers - simple pH measurement
  • Universal and special indicator papers supplied in plastic reels that ensure protection against external influences .
  • Always ready for use .
  • The reaction color of the paper is compared with the color scale and read the pH . Allows a very simple pH measurement .
Specifications
Certifications/Compliance ISO 9001
pH Range 9 to 13
Format Roll
Graduations 9.0, 9.5, 10.0, 10.5, 11.0, 11.5, 12.0, 12.5, 13.0
Quantity 1 Each
Type pH Paper