Learn More
Macherey-Nagel™ pH Paper
Product Code. 10570022 Shop All Macherey-Nagel Products
Change view
Click to view available options
pH Range:
0.5 to 5.5
1 to 11
1 to 14
3.8 to 5.8
4 to 7
5.4 to 7
6.4 to 8
7.2 to 9.7
9 to 13
Unit Size:
5m
Each
Pack of 5
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. pH Range Unit Size
10570022 6.4 to 8 5m
10797941 1 to 14 5m
10098250 0.5 to 5.5 Each
10464491 1 to 11 Each
10323361 3.8 to 5.8 5m
10539641 4 to 7 Pack of 5
10549641 5.4 to 7 Pack of 5
10613141 7.2 to 9.7 5m
10527341 9 to 13 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 10570022 Supplier Macherey-Nagel™ Supplier No. 90210
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Measure pH simply and easily. Macherey-Nagel™ pH paper comes in a circular format with a wide variety of pH ranges.
  • Universal and special indicator papers - simple pH measurement
  • Universal and special indicator papers supplied in plastic reels that ensure protection against external influences .
  • Always ready for use .
  • The reaction color of the paper is compared with the color scale and read the pH . Allows a very simple pH measurement .
Specifications
Certifications/Compliance ISO 9001
pH Range 6.4 to 8
Format Roll
Graduations <6.4, 6.4, 6.6, 6.8, 7.0, 7.2, 7.4, 7.6, 7.8, 8.0, >8.0
Quantity 1 Each
Type pH Paper