missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHACS Antibody (1D2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00084680-M02
This item is not returnable.
View return policy
Description
PHACS Monoclonal antibody specifically detects PHACS in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PHACS | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| 1-aminocyclopropane-1-carboxylate synthase homolog(Arabidopsis)(non-functional), 1-aminocyclopropane-1-carboxylate synthase-like protein 1, PHACSACSACC synthase-like protein 1 | |
| PHACS (NP_115981, 1 a.a. ∽ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MFTLPQKDFRAPTTCLGPTCMQDLGSSHGEDLEGECSRKLDQKLPELRGVGDPAMISSDTSYLSSRGRMIKWFWDSAEEGYRTYHMDEYDEDKNPSGIIN | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1D2 | |
| Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| NP_115981 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 84680 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction