missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHACTR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56787
This item is not returnable.
View return policy
Description
PHACTR1 Polyclonal specifically detects PHACTR1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Polyclonal | |
| dJ257A7.2, KIAA1733MGC126577, MGC126575, phosphatase and actin regulator 1, RPEL, RPEL repeat containing 1, RPEL1 | |
| Rabbit | |
| 100 μL | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| PHACTR1 | |
| Affinity purified | |
| Mouse, Rat, Bovine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction