missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHACTR3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19790
This item is not returnable.
View return policy
Description
PHACTR3 Polyclonal specifically detects PHACTR3 in Human samples. It is validated for Western Blot.
Specifications
| PHACTR3 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 20% Glycerol (pH7) with 0.025% Proclin 300 | |
| C20orf101, chromosome 20 open reading frame 101, H17739, MGC117178, phosphatase and actin regulator 3, scaffold-associated PP1 inhibiting protein, Scaffold-associated PP1-inhibiting protein, SCAPIN1, Scapinin | |
| Rabbit | |
| 63 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Species reactivity based on sequence identity: Mouse (82%), Rat (81%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.96mg/mL | |
| Western Blot 1:5000-1:100000 | |
| Q96KR7 | |
| PHACTR3 | |
| Recombinant protein encompassing a sequence within the center region of human PHACTR3. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 116154 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction