Learn More
Magle Life Sciences™ Phadebas™ Saliva Amylase-detection Forensic Paper
Product Code. p-8014740 Shop All Magle Life Sciences Products
Click to view available options
Dimensions (L x W):
300 x 195 mm
395mm x 15m roll
Unit Size:
Each
Pack of 12
Product Code. 12895552
Brand: Magle Life Sciences™ 1314
Only null left
Add to basket
Add to basket
Used for presumptive testing of suspicious stains. Phadebas™ Saliva Amylase-detection Forensic Paper is based on the Phadebas Amylase Test tablets but follows a separate method developed by the Forensic Biology labs.
Pre-manufactured papers;
  • Are applicable directly from package;
  • Include Batch number and Expiry date;
  • Are easy to handle and use;
  • Have negligible batch variations;
  • Give no difference between labs and consequently have higher credibility as evidence material
Specifications
Dimensions (L x W) 300 x 195 mm