missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phafin 1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17187468 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17187468 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17187468 Supplier Invitrogen™ Supplier No. PA5116236

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Phafin 1 Polyclonal Antibody for Western Blot

APPD (Apoptosis-inducing protein D; phafin 1) is predicted through gene annotation and contains pleckstrin homology domain. It may binds to two Zn++ ions through its FYVE zinc finger domain. Biological function of this protein has not been clearly mapped yet.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phafin 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PLEKHF1
Gene Accession No. Q96S99
Gene Alias 1810013P09Rik; AI037048; Apoptosis-inducing protein; apoptosis-inducing protein D; APPD; Lapf; lysosome-associated apoptosis-inducing protein containing PH and FYVE domains; PH and FYVE domain-containing protein 1; PH domain-containing family F member 1; phafin 1; PHAFIN1; phafin-1; pleckstrin homology and FYVE domain containing 1; pleckstrin homology domain containing, family F (with FYVE domain) member 1; pleckstrin homology domain-containing family F member 1; Plekhf1; SGP-1; ZFYVE15; Zinc finger FYVE domain-containing protein 15
Gene Symbols PLEKHF1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 51-79 amino acids from the N-terminal region of human PLEKHF1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79156
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.