missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phakinin Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 196.00 - € 468.00
Specifications
| Antigen | phakinin |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
phakinin Polyclonal antibody specifically detects phakinin in Mouse samples. It is validated for Western BlotSpecifications
| phakinin | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytoskeleton Markers, Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 8419 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| beaded filament structural protein 2, phakinin, CP47, CP49, Cytoskeletal protein, 49-kD, MGC142078, phakinin | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 116-415 of human BFSP2 (NP_003562.1). VEYMAKVHALEQVSQELETQLRMHLESKATRSGNWGALRASWASSCQQVGEAVLENARLMLQTETIQAGADDFKERYENEQPFRKAAEEEINSLYKVIDEANLTKMDLESQIESLKEELGSLSRNYEEDVKLLHKQLAGCELEQMDAPIGTGLDDILETIRIQWERDVEKNRVEAGALLQAKQQAEVAHMSQTQEEKLAAALRVELHNTSCQVQSLQAETESLRALKRGLENTLHDAKHWHDMELQNLGAVVGRLEAELREIRAEAEQQQQERAHLLARKCQLQKDVASYHALLDREESG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title