missing translation for 'onlineSavingsMsg'
Learn More
Learn More
phakinin Antibody (OTI4E7), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73365DL594
This item is not returnable.
View return policy
Description
phakinin Monoclonal antibody specifically detects phakinin in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| phakinin | |
| Monoclonal | |
| DyLight 594 | |
| beaded filament structural protein 2, phakinin, CP47, CP49, Cytoskeletal protein, 49-kD, MGC142078, phakinin | |
| Recombinant protein fragment corresponding to amino acids 184-415 of human phakinin (NP_003562) produced in E.coli. | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Neuroscience | |
| 8419 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI4E7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction