Learn More
Masterflex™ L/S™ PharMed™ BPT Spooled Pump Tubing
Product Code. p-8015417
Click to view available options
Tubing Size:
14
15
16
17
18
24
35
36
Unit Size:
Each
Product Code. 11745248
Brand: Masterflex™ 9568715
Only null left
Add to basket
Add to basket
Use this spooled tubing for cell and tissue culture work and pharmaceutical uses. Masterflex™ L/S™ PharMed™ BPT Spooled Pump Tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport.
  • Opaque beige spooled pump tubing requires less storage space than the equivalent amount of individual tubing coils
  • Heat-sealable, bondable and formable
  • Ideal for light-sensitive samples
  • Nontoxic, nonhemolytic, extremely low permeability
  • Resists acids, alkalis, oxidizing agents and animal and vegetable oils
  • Reduce scrap, cut only as much as you need
  • Ensure that you always have enough tubing on hand
Nearly any Masterflex pump tubing formulation can be spooled. Call our Application Specialists for a quote or to place a special order
Specifications
Autoclavable Autoclavable
Diameter (Metric) Inner 4.8 mm
Material PharMed BPT
Overall Length (English) 400 ft.
Overall Length (Metric) 12,190 cm
Min. Operating Temperature (English) -60°F
Min. Operating Temperature (Metric) -51°C
Tubing Size 15
Flow Rate 1100 mL/min.
Hose Barb 3/16 in.
Height (Metric) 8.8 m Water
Height (English) 29 ft. Water
Color Opaque Beige
Maximum Pressure 40 psi (2.7 bar)
Vacuum Level 660 mm Hg (26 in. Hg)
Max. Operating Temperature (English) 270°F
Max. Operating Temperature (Metric) 132°C
Product Type Pump Tubing