missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHAX Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22268C
This item is not returnable.
View return policy
Description
PHAX Polyclonal antibody specifically detects PHAX in Human, Mouse samples. It is validated for Western Blot
Specifications
| PHAX | |
| Polyclonal | |
| Western Blot | |
| FLJ13193, phosphorylated adapter RNA export protein, phosphorylated adaptor for RNA export, RNA U small nuclear RNA export adapter protein, RNA U, small nuclear RNA export adapter (phosphorylation regulated), RNA U, small nuclear RNA export adaptor (phosphorylation regulated), RNUXA | |
| The immunogen this antibody was made to, maps to a region between residue 344 and 394 of human Phosphorylated Adaptor for RNA Export using the numbering given in entry NP_115553.2 (GeneID 51808). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 51808 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction