Learn More
Invitrogen™ PHC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120956
Description
Positive test controls include: A-549, Mouse kidney. The target is usually found in the following locations: Nucleus. Immunogen sequence: FPVGCSQLLK ESEKPLQTGL PTGLTENQSG GPLGVDSPSA ELDKKANLLK CEYCGKYAPA EQFRGSKRFC SMTCAKRYNV SCSHQFRLKR KKMKEFQEAN YARVRRRGPR RSSSDIARAK IQGKCHRGQE DSSRGSDNSS YDEALSPTSP GPLSVRAGHG ERDLGNPNTA PPTPELHGIN PVFLSSNPSR W.
Polycomb group (PcG) proteins assemble into multimeric protein complexes, which are involved in maintaining the transcriptional repressive state of genes throughout development. PHC1 (polyhomeotic homolog 1), also known as EDR1, HPH1 or RAE28, is a 1,004 amino acid nuclear protein that is a component of the PcG multiprotein PRC1 complex. Specifically, the PcG PRC1 complex modifies histones, remodels chromatin and mediates monoubiquination of Histone H2A. Other constituent proteins involved in the PcG PRC1 complex are Mel-18, Bmi-1, M33, MPc2, MPc3, RING1, Ring1b, as well as several others. Existing as a homodimer, PHC1 contains one FCS-type zinc finger and a SAM (sterile alpha motif) domain. PHC1 is encoded by a gene located on human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome.
Specifications
| PHC1 | |
| Polyclonal | |
| Unconjugated | |
| PHC1 | |
| AW557034; DKFZp686A1782; early development regulator 1 (homolog of polyhomeotic 1); early development regulator1 (homolog of polyhomeotic 1); early development regulatory protein 1; Edr; Edr1; hPH1; MCPH11; Mph1; PH1; Phc1; polyhomeotic homolog 1; polyhomeotic homolog 1 (Drosophila); polyhomeotic-like 1; polyhomeotic-like 1 (Drosophila); polyhomeotic-like protein 1; RAE28; RAE-28 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13619, 1911, 312690 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P78364, Q64028 | |
| PHC1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 750-940 of human PHC1 (NP_004417.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.