missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589939
This item is not returnable.
View return policy
Description
Immunogen sequence: HFLPSEPTKW NVEDVYEFIR SLPGCQEIAE EFRAQEIDGQ ALLLLKEDHL MSAMNIKLGP ALKIYARISM LKDS; Positive Samples: DU145, HT-29, HeLa, Jurkat, 293T; Cellular Location: Nucleus.
In Drosophila melanogaster, the 'Polycomb' group (PcG) of genes are part of a cellular memory system that is responsible for the stable inheritance of gene activity. PcG proteins form a large multimeric, chromatin-associated protein complex. The protein encoded by this gene has homology to the Drosophila PcG protein 'polyhomeotic' (Ph) and is known to heterodimerize with EDR1 and colocalize with BMI1 in interphase nuclei of human cells. The specific function in human cells has not yet been determined. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| PHC2 | |
| Polyclonal | |
| Unconjugated | |
| Phc2 | |
| A3galt2; AA415044; D130050K19Rik; D4Ertd810e; early development regulator 2; early development regulator 2 (homolog of polyhomeotic 2); early development regulatory protein 2; Edr2; hPH2; mPH2; p36; Ph2; Phc2; polyhomeotic 2; polyhomeotic 2 protein; polyhomeotic homolog 2; polyhomeotic homolog 2 (Drosophila); polyhomeotic-like 2; polyhomeotic-like 2 (Drosophila); polyhomeotic-like protein 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1912 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.32 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8IXK0 | |
| Phc2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 250-323 of human PHC2 (NP_0044182). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion