missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHC3 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82020AF647
This item is not returnable.
View return policy
Description
PHC3 Polyclonal antibody specifically detects PHC3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| PHC3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| Alexa Fluor 647 | |
| DKFZp313K1221, early development regulator 3, Early development regulatory protein 3, EDR3FLJ12729, FLJ12967, Homolog of polyhomeotic 3, HPH3, MGC88144, PH3, polyhomeotic homolog 3 (Drosophila), polyhomeotic like 3, polyhomeotic like 3 (Drosophila), polyhomeotic-like protein 3 | |
| PHC3 antibody was raised against a 19 amino acid peptide near the amino terminus of human PHC3. The immunogen is located within amino acids 240 - 290 of PHC3. Amino Acid Squence: KISQRDPSPESNKKGESPS | |
| 0.1 mL | |
| Chromatin Modifiers | |
| 80012 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction