missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHC3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94030-0.1ml
This item is not returnable.
View return policy
Description
PHC3 Polyclonal antibody specifically detects PHC3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| PHC3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| DKFZp313K1221, early development regulator 3, Early development regulatory protein 3, EDR3FLJ12729, FLJ12967, Homolog of polyhomeotic 3, HPH3, MGC88144, PH3, polyhomeotic homolog 3 (Drosophila), polyhomeotic like 3, polyhomeotic like 3 (Drosophila), polyhomeotic-like protein 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 100-300 of human PHC3 (NP_079223.3). LSSSQLQSLAAVQASLSSGRPSTSPTGSVTQQSSMSQTSINLSTSPTPAQLISRSQASSSTSGSITQQTMLLGSTSPTLTASQAQMYLRAQMLIFTPATTVAAVQSDIPVVSSSSSSSCQSAATQVQNLTLRSQKLGVLSSSQNGPPKSTSQTQSLTICHNKTTVTSSKISQRDPSPESNKKGESPSLESRSTAVTRTSSI | |
| 0.1 mL | |
| Chromatin Modifiers | |
| 80012 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction