missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHC3 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-77312H
This item is not returnable.
View return policy
Description
PHC3 Polyclonal antibody specifically detects PHC3 in Human samples. It is validated for Western Blot
Specifications
| PHC3 | |
| Polyclonal | |
| Western Blot | |
| DKFZp313K1221, early development regulator 3, Early development regulatory protein 3, EDR3FLJ12729, FLJ12967, Homolog of polyhomeotic 3, HPH3, MGC88144, PH3, polyhomeotic homolog 3 (Drosophila), polyhomeotic like 3, polyhomeotic like 3 (Drosophila), polyhomeotic-like protein 3 | |
| The immunogen recognized by this antibody maps to a region between residue 700 and 750 of human polyhomeotic like 3 using the numbering given in entry AAI31773.1 (GeneID 80012). | |
| 0.1 mL | |
| Chromatin Modifiers | |
| 80012 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction