Learn More
PHCCC, MedChemExpress
Product Code. 16485737
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485737 5 mg 5mg
16482576 25 mg 25mg
16474306 100 mg 100mg
16472998 10 mg 10mg
16469566 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16485737 Supplier Medchem Express Supplier No. HY100409
MedChemExpress PHCCC is a Group I mGluR antagonist with an IC50 of 3 μM. PHCCC is a selective positive modulator of mGlu4 receptor. Antiparkinsonian effect.
PHCCC is a Group I mGluR antagonist with an IC50 of 3 μM. PHCCC is a selective positive modulator of mGlu4 receptor. Antiparkinsonian effect.
Specifications
Chemical Name or Material PHCCC
CAS 179068-02-1
Color White
Purity Grade Notes Research
Molecular Formula C17H14N2O3
Quantity 5 mg
Solubility Information DMSO : 12.5 mg/mL (42.47 mM; Need ultrasonic)
SMILES O=C(C12C(C2)/C(C3=CC=CC=C3O1)=N\O)NC4=CC=CC=C4
Molecular Weight (g/mol) 294.3
Formula Weight 294.3
Percent Purity 99.96%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only