missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHD4/HIF Prolyl Hydroxylase 4 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-295DL594
This item is not returnable.
View return policy
Description
PHD4/HIF Prolyl Hydroxylase 4 Polyclonal antibody specifically detects PHD4/HIF Prolyl Hydroxylase 4 in Human, Rat, Mouse (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Electron Microscopy
Specifications
| PHD4/HIF Prolyl Hydroxylase 4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat, Mouse (Negative) | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Electron Microscopy | |
| DyLight 594 | |
| EC 1.14.11.-, EGLN4, HIFPH4, HIF-PH4, HIF-prolyl hydroxylase 4, HPH-4, hypoxia-inducible factor prolyl 4-hydroxylase, Hypoxia-inducible factor prolyl hydroxylase 4, P4H with transmembrane domain, P4H-TMFLJ20262, PH-4, PH4 hypoxia inducible factor prolyl 4 hydroxylase, PHD4, proline 4-hydroxylase, prolyl 4-hydroxylase, transmembrane (endoplasmic reticulum), Prolyl hydroxlase domain-containing 4, transmembrane prolyl 4-hydroxylase | |
| The epitope recognized by this antibody maps to a region between residue 450 and the C-terminus (residue 502) of human PHD4/HIF Prolyl Hydroxylase 4 using the numbering given in entry NP_808808.1 (GeneID 54681). | |
| 0.1 mL | |
| Cancer, Hypoxia, Lipid and Metabolism | |
| 54681 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction