missing translation for 'onlineSavingsMsg'
Learn More

Phenethylamine-d4, TRC

Product Code. 30413271
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413271 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30413271 Supplier Toronto Research Chemicals Supplier No. TRCP321336100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenethylamine-d4
CAS 87620-08-4
Molecular Formula C8 2H4 H7 N
Quantity 100 mg
Synonym Benzeneethan-d4-amine (9CI','ACI), Benzeneethan-1,1,2,2-d4-amine, Phenethylamine-d4, 2-Phenylethyl-1,1,2,2-d4-amine, 2-Phenylethylamine-1,1,2,2-D4
InChI Formula InChI=1S/C8H11N/c9-7-6-8-4-2-1-3-5-8/h1-5H,6-7,9H2/i6D2,7D2
SMILES [2H]C([2H])(N)C([2H])([2H])c1ccccc1
IUPAC Name 1,1,2,2-tetradeuterio-2-phenylethanamine
Molecular Weight (g/mol) 125.2
Formula Weight 125.1143
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.