Learn More
Phenindione, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Phenindione
Specifications
Specifications
| Chemical Name or Material | Phenindione |
| CAS | 83-12-5 |
| Analyte or Component Names | Phenindione |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H10 O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 1H-Indene-1,3(2H)-dione, 2-phenyl-, 1,3-Indandione, 2-phenyl- (6CI,8CI), 2-Phenyl-1,3-diketohydrindene, 2-Phenyl-1,3-indandione, 2-Phenyl-1,3-indanedione, 2-Phenyl-1H-indene-1,3(2H)-dione, 2-Phenylindandione, Athrombon, Bindan, Cronodione, Danedion, Danilon, Danilone, Diadilan, Dindevan, Dineval, Diophindane, Emandion, Emandione, Fenhydren, Fenilin, Fenindion, Hedulin, Hemolidione, Indema, Indion, Indon, NSC 41693, PID, Phenhydren, Phenillin, Phenindione, Phenylen, Phenylin, Phenylindione, Pindione, Rectadione, Theradione, Thrombasal, Tromazal, Trombol |
| InChI Formula | InChI=1S/C15H10O2/c16-14-11-8-4-5-9-12(11)15(17)13(14)10-6-2-1-3-7-10/h1-9,13H |
| SMILES | O=C1C(C(=O)c2ccccc12)c3ccccc3 |
| IUPAC Name | 2-phenylindene-1,3-dione |
| Show More |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur