missing translation for 'onlineSavingsMsg'
Learn More

Phenothrin (>85%), TRC

Product Code. 30409668
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30409668 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409668 Supplier Toronto Research Chemicals Supplier No. TRCP31809510G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenothrin
CAS 26002-80-2
Molecular Formula C23 H26 O3
Quantity 10 g
Synonym Cyclopropanecarboxylic acid','2,2-dimethyl-3-(2-methyl-1-propen-1-yl)-','(3-phenoxyphenyl)methyl ester, Cyclopropanecarboxylic acid','2,2-dimethyl-3-(2-methyl-1-propenyl)-','(3-phenoxyphenyl)methyl ester (9CI), Cyclopropanecarboxylic acid','2,2-dimethyl-3-(2-methylpropenyl)-','m-phenoxybenzyl ester (8CI), Benzyl alcohol','m-phenoxy-','2,2-dimethyl-3-(2-methylpropenyl)cyclopropanecarboxylate (8CI), 3-Phenoxybenzyl chrysanthemate, Anchimanaito 20S, Duet, Multicide 2154, OMS 1809, OMS 1810, PT 515, Phenothrin, Phenoxythrin, Pibutin, S 2539, S 2539 (pesticide), m-Phenoxybenzyl 2,2-dimethyl-3-(2-methylpropenyl)cyclopropanecarboxylate
InChI Formula InChI=1S/C23H26O3/c1-16(2)13-20-21(23(20,3)4)22(24)25-15-17-9-8-12-19(14-17)26-18-10-6-5-7-11-18/h5-14,20-21H,15H2,1-4H3
SMILES CC(=CC1C(C(=O)OCc2cccc(Oc3ccccc3)c2)C1(C)C)C
IUPAC Name (3-phenoxyphenyl)methyl 2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
Molecular Weight (g/mol) 350.45
Formula Weight 350.1882
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.