missing translation for 'onlineSavingsMsg'
Learn More

Phenothrin, MedChemExpress

Product Code. 16468586
Encompass
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468586 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468586 Supplier Medchem Express Supplier No. HYB1072

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Phenothrin is a synthetic pyrethroid that kills adult fleas and ticks.

Phenothrin is a synthetic pyrethroid that kills adult fleas and ticks.

Specifications

Chemical Name or Material Phenothrin
CAS 26002-80-2
Color Colorless
Purity Grade Notes Research
Molecular Formula C23H26O3
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (285.35 mM; Need ultrasonic)
SMILES O=C(C1C(C)(C)C1/C=C(C)\C)OCC2=CC=CC(OC3=CC=CC=C3)=C2
Molecular Weight (g/mol) 350.45
Formula Weight 350.45
Percent Purity 94.0%
Grade Research
Physical Form Liquid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.