Learn More
Phenprocoumon, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Phenprocoumon
Specifications
Specifications
| Chemical Name or Material | Phenprocoumon |
| CAS | 435-97-2 |
| Analyte or Component Names | Phenprocoumon |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H16 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 3-(alpha-Ethylbenzyl)-4-hydroxycoumarin, (+/-)-Phenprocoumon, 3-(1-Phenylpropyl)-4-hydroxycoumarin, 3-(alpha-Phenylpropyl)-4-hydroxycoumarin, 4-Hydroxy-2-oxo-3-(1-phenylpropyl)-2H-chromene, DL-3-(alpha-Ethylbenzyl)-4-hydroxycoumarin, Falithrom, Fencumar, Liquamar, Marcoumar, Marcumar, Phenprocoumarol, Phenprocoumarole, Phenprocoumon |
| InChI Formula | InChI=1S/C18H16O3/c1-2-13(12-8-4-3-5-9-12)16-17(19)14-10-6-7-11-15(14)21-18(16)20/h3-11,13,19H,2H2,1H3 |
| SMILES | CCC(C1=C(O)c2ccccc2OC1=O)c3ccccc3 |
| IUPAC Name | 4-hydroxy-3-(1-phenylpropyl)chromen-2-one |
| Show More |