Learn More
Phenserine, TRC
Specifications
Specifications
| Chemical Name or Material | Phenserine |
| CAS | 101246-66-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C20H23N3O2 |
| Quantity | 50 mg |
| Synonym | (-)-Eseroline Phenylcarbamate, (-)-N-Phenylcarbamoyleseroline, (-)-Phenserine, (3aS-cis)-1,2,3,3a,8,8a-Hexahydro-1,3a,8-trimethyl-pyrrolo[2,3-b]indol-5-ol Phenylcarbamate (Ester), (3aS,8aR)-1,2,3,3a,8,8a-Hexahydro-1,3a,8-trimethyl-pyrrolo[2,3-b]indol-5-ol (N-Phenylcarbamate) |
| InChI Formula | InChI=1S/C20H23N3O2/c1-20-11-12-22(2)18(20)23(3)17-10-9-15(13-16(17)20)25-19(24)21-14-7-5-4-6-8-14/h4-10,13,18H,11-12H2,1-3H3,(H,21,24)/t18-,20+/m1/s1 |
| SMILES | CN1CC[C@]2(C)[C@H]1N(C)c3ccc(OC(=O)Nc4ccccc4)cc23 |
| IUPAC Name | [(3aR,8bS)-3,4,8b-trimethyl-2,3a-dihydro-1H-pyrrolo[2,3-b]indol-7-yl] N-phenylcarbamate |
| Molecular Weight (g/mol) | 337.42 |
| Show More |