Learn More
Phenyl 1-Thio-Beta-D-galactoside, TRC
Product Code. 30422287
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30422287 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30422287 Supplier Toronto Research Chemicals Supplier No. TRCP3369801G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Phenyl 1-Thio-Beta-D-galactoside
CAS 16758-34-2
Molecular Formula C12 H16 O5 S
Quantity 1 g
InChI Formula InChI=1S/C12H16O5S/c13-6-8-9(14)10(15)11(16)12(17-8)18-7-4-2-1-3-5-7/h1-5,8-16H,6H2/t8-,9+,10+,11-,12+/m1/s1
SMILES OC[C@H]1O[C@@H](Sc2ccccc2)[C@H](O)[C@@H](O)[C@H]1O
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-phenylsulfanyloxane-3,4,5-triol
Molecular Weight (g/mol) 272.32
Formula Weight 272.0718