missing translation for 'onlineSavingsMsg'
Learn More

Phenyl nicotinate, 99%, Thermo Scientific™

Product Code. 11425856
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425856 100 g 100g
11405856 5 g 5g
11415856 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425856 Supplier Thermo Scientific Alfa Aesar Supplier No. B22219.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
In nicotinic acid analogs assayed against tri-o-cresyl phosphate-induced ataxia in hens Phenyl nicotinate provided partial relief of the syndrome.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3468-53-9
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD00023581
InChI Key QEJPOSAIULNDLU-UHFFFAOYSA-N
Synonym phenyl nicotinate, 3-pyridinecarboxylic acid, phenyl ester, phenylnicotinate, phenyl 3-pyridinecarboxylate, nicotinic acid, phenyl ester, unii-vrd2l6rm16, vrd2l6rm16, acmc-209i9k, nicotinic acid phenyl ester, qejposaiulndlu-uhfffaoysa
PubChem CID 77018
IUPAC Name phenyl pyridine-3-carboxylate
SMILES C1=CC=C(C=C1)OC(=O)C2=CN=CC=C2

Specifications

Melting Point 70°C to 74°C
Boiling Point 186°C to 188°C (20 mmHg)
Quantity 100 g
Beilstein 140968
Formula Weight 199.21
Percent Purity 99%
Chemical Name or Material Phenyl nicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.