Learn More
Phenyl selenocyanate, 98%
Product Code. p-7048072
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11359026
Brand: Thermo Scientific Alfa Aesar L20194.03
Request Bulk or Custom Format
CAS: 2179-79-5 | C7H5NSe | 182.095 g/mol
Phenyl selenocyanate is used as a specific source of nucleophilic and electrophilic selenium species in synthetic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phenyl selenocyanate is used as a specific source of nucleophilic and electrophilic selenium species in synthetic chemistry.
Solubility
Miscible with organic solvents, terahydro furan, dichloromethane and acetonitrile.
Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2179-79-5
Molecular Formula C7H5NSe
Molecular Weight (g/mol) 182.095
MDL Number MFCD00216944
InChI Key NODWRXQVQYOJGN-UHFFFAOYSA-N
Synonym phenylselenocyanate, phenylselanyl formonitrile, selenocyanic acid, phenyl ester, acmc-1cgab, phenyl selenocyanate, selenocyanic acid phenyl ester, 2-phenyl-2-selenaethanenitrile
PubChem CID 555340
IUPAC Name phenyl selenocyanate
SMILES C1=CC=C(C=C1)[Se]C#N
Specifications
Density 1.484
Boiling Point 116°C to 117°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.603
Quantity 1 g
UN Number UN3440
Beilstein 1617370
Solubility Information Miscible with organic solvents,terahydro furan,dichloromethane and acetonitrile.
Formula Weight 182.08
Percent Purity 98%
Chemical Name or Material Phenyl selenocyanate