missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetic acid, 98.5%

Product Code. 10627294 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Unit Size:
2.5kg
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10627294 2.5 kg 2.5kg
10795421 5 g 5g
10316970 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 10627294 Supplier Thermo Scientific Acros Supplier No. 130280025
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

CAS: 103-82-2 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-82-2
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
InChI Key WLJVXDMOQOGPHL-UHFFFAOYSA-N
IUPAC Name 2-phenylacetic acid
SMILES OC(=O)CC1=CC=CC=C1

Specifications

Melting Point 75.0°C to 78.0°C
Density 1.0800 g/mL
Boiling Point 265.0°C
Flash Point 132°C
Assay Percent Range 98.5%
Packaging Plastic bottle
Linear Formula C6H5CH2CO2H
Quantity 2.5 kg
Beilstein 09, 431
Merck Index 15, 7380
Specific Gravity 1.08
Solubility Information Solubility aq. soln.: 15 g/L (20°C). Other solubilities: freely soluble in hot water, soluble in ether, acetone, 186 g/100 mL ethanol (20°C), 4.422 mol.s/l in chloroform (25°C), 1.842 mol.s/l in carbon tetrachloride
Formula Weight 136.15
Chemical Name or Material Phenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • Phenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.