Learn More
Phenylacetylene-d6, CDN
Specifications
Specifications
| Chemical Name or Material | Ethynylbenzene-D6 |
| CAS | 25837-47-2 |
| Molecular Formula | C8 D6 |
| Quantity | 0.25 g |
| Synonym | Benzene-1,2,3,4,5-d5, 6-(ethynyl-2-d)- (ACI), 6-(Ethynyl-2-d)benzene-1,2,3,4,5-d5 (ACI), Benzene-d5, ethynyl-d- (8 CI, 9 CI), Phenyl-d5-acetylene-d, Ethynylbenzene-d6, Phenylacetylene-d6 |
| InChI Formula | InChI=1 S/C8H6/c1-2-8-6-4-3-5-7-8/h1,3-7 H/i1D,3 D,4 D,5 D,6 D,7 D |
| SMILES | [2 H]C#Cc1c([2 H])c([2 H])c([2 H])c([2 H])c1[2 H] |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-(2-deuterioethynyl)benzene |
| Molecular Weight (g/mol) | 108.1702 |
| Formula Weight | 108.0846 g/mol |
| Show More |