missing translation for 'onlineSavingsMsg'
Learn More

Phenylacetylglutamine, MedChemExpress

Product Code. 16485457
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485457 50 mg 50mg
16430489 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16485457 Supplier Medchem Express Supplier No. HYW050026

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Phenylacetylglutamine is a colonic microbial metabolite from amino acid fermentation.

Phenylacetylglutamine is a colonic microbial metabolite from amino acid fermentation.

Specifications

Chemical Name or Material Phenylacetylglutamine
CAS 28047-15-6
Color White
Purity Grade Notes Research
Molecular Formula C13H16N2O4
Synonym NSC 203800 Phenylacetyl-L-glutamine
Solubility Information DMSO : 100 mg/mL (378.39 mM; Need ultrasonic) ∣H2O : 25 mg/mL (94.60 mM; Need ultrasonic)
SMILES O=C([C@H](CCC(N)=O)NC(CC1=CC=CC=C1)=O)O
Molecular Weight (g/mol) 264.28
Quantity 50 mg
Formula Weight 264.28
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.