missing translation for 'onlineSavingsMsg'
Learn More

Phenylephrine Maleic Acid Adduct Hydrochloride (2-[6-(3-Hydroxyphenyl)-4-methyl-2-oxomorpholin-3-yl]acetic Acid Hydrochloride), Mikromol™

Product Code. 17910576
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17910576 25 mg 25mg
17950646 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17910576 Supplier Mikromol Supplier No. MM0081.120025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Phenylephrine Maleic Acid Adduct Hydrochloride (2-[6-(3-Hydroxyphenyl)-4-methyl-2-oxomorpholin-3-yl]acetic Acid Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[6-(3-Hydroxyphenyl)-4-methyl-2-oxomorpholin-3-yl]acetic Acid Hydrochloride
CAS 2575516-52-6
Analyte or Component Names 2-[6-(3-Hydroxyphenyl)-4-methyl-2-oxomorpholin-3-yl]acetic Acid Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C13 H15 N O5 . Cl H
Certifications/Compliance ISO 17025
Synonym 3-(Carboxymethyl)-6-(3-hydroxyphenyl)-4-methyl-2-oxomorpholin-4-ium Chloride, PEMA (phenylephrine maleic acid adduct)
InChI Formula InChI=1S/C13H15NO5.ClH/c1-14-7-11(8-3-2-4-9(15)5-8)19-13(18)10(14)6-12(16)17;/h2-5,10-11,15H,6-7H2,1H3,(H,16,17);1H
SMILES Cl.CN1CC(OC(=O)C1CC(=O)O)c2cccc(O)c2
IUPAC Name 2-[6-(3-hydroxyphenyl)-4-methyl-2-oxomorpholin-3-yl]acetic acid;hydrochloride
Molecular Weight (g/mol) 301.72
Quantity 25 mg
Formula Weight 301.0717 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.