Learn More
Invitrogen™ PHEX Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143870
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: rat ovary tissue, rat kidney tissue, mouse ovary tissue, mouse kidney tissue, human HEK293 whole cell, monkey kidney tissue, rat NRK whole cell, monkey COS-7 whole cell. IHC: human lung cancer tissue, human ovarian cancer tissue. Flow: U87 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Peptidase that cleaves SIBLING (small integrin-binding ligand, N-linked glycoprotein)-derived ASARM peptides, thus regulating their biological activity. Cleaves ASARM peptides between Ser and Glu or Asp residues. Regulates osteogenic cell differentiation and bone mineralization through the cleavage of the MEPE-derived ASARM peptide. Promotes dentin mineralization and renal phosphate reabsorption by cleaving DMP1- and MEPE-derived ASARM peptides. Inhibits the cleavage of MEPE by CTSB/cathepsin B thus preventing MEPE degradation.
Specifications
| PHEX | |
| Polyclonal | |
| Unconjugated | |
| PHEX | |
| Gy; HPDR; HPDR1; HYP; HYP1; LXHR; metalloendopeptidase homolog PEX; Metalloendopeptidase homolog PEX (Phosphate regulating neutral endopeptidase) (X-linked hypophosphatemia protein) (HYP) (Vitamin D-resistant hypophosphatemic rickets protein); PEX; PHEX; phosphate regulating endopeptidase homolog, X-linked; phosphate regulating gene with homologies to endopeptidases on the X chromosome (hypophosphatemia, vitamin D resistant rickets); phosphate regulating neutral endopeptidase; Phosphate regulating neutral endopeptidase on the X chromosome (X-linked hypophosphatemia XLH); phosphate-regulating neutral endopeptidase; Phosphate-regulating neutral endopeptidase PHEX; Vitamin D-resistant hypophosphatemic rickets protein; XLH; X-linked hypophosphatemia protein; X-linked phosphate regulating endopeptidase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18675, 25512, 5251 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P70669, P78562 | |
| PHEX | |
| E.coli-derived human PHEX recombinant protein (Position: E145-N718). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.